Anti-RPS19BP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315981EA 552 EUR
ORIGTA315981
Anti-RPS19BP1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: RPS19BP1 (ribosomal protein S19 binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR