Anti-SLC25A21 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315965EA 552 EUR
ORIGTA315965
Anti-SLC25A21 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: SLC25A21 (solute carrier family 25 (mitochondrial oxoadipate carrier), member 21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR