Anti-ATG16L2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315964EA 552 EUR
ORIGTA315964
Anti-ATG16L2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: ATG16L2 (autophagy related 16-like 2 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR