Anti-RELT Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315947EA 564 EUR
ORIGTA315947
Anti-RELT Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: RELT (RELT tumor necrosis factor receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR