Anti-CEP290 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315824EA 564 EUR
ORIGTA315824
Anti-CEP290 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CEP290 (centrosomal protein 290kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR