Anti-TRAPPC6A Rabbit Polyclonal Antibody

Proveedor: OriGene
TA315773
ORIGTA315773EA 543 EUR
ORIGTA315773
Anti-TRAPPC6A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: TRAPPC6A (trafficking protein particle complex 6A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR