Anti-MRPL32 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315759EA 552 EUR
ORIGTA315759
Anti-MRPL32 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: MRPL32 (mitochondrial ribosomal protein L32)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR