Anti-CREB3L2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315744EA 532 EUR
ORIGTA315744
Anti-CREB3L2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: CREB3L2 (cAMP responsive element binding protein 3-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR