Anti-TMBIM1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315717EA 552 EUR
ORIGTA315717
Anti-TMBIM1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: TMBIM1 (transmembrane BAX inhibitor motif containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR