Anti-SENP7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315656EA 552 EUR
ORIGTA315656
Anti-SENP7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: SENP7 (SUMO1/sentrin specific peptidase 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR