Anti-CLN6 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315557EA 564 EUR
ORIGTA315557
Anti-CLN6 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CLN6 (ceroid-lipofuscinosis, neuronal 6, late infantile, variant)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR