Anti-GNL3L Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315530EA 543 EUR
ORIGTA315530
Anti-GNL3L Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: GNL3L (guanine nucleotide binding protein-like 3 (nucleolar)-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR