Anti-CALML5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315504EA 532 EUR
ORIGTA315504
Anti-CALML5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: CALML5 (calmodulin-like 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR