Anti-CYP39A1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315463EA 552 EUR
ORIGTA315463
Anti-CYP39A1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: CYP39A1 (cytochrome P450, family 39, subfamily A, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR