Anti-IL17RA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315273EA 552 EUR
ORIGTA315273
Anti-IL17RA Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:60000

Type: Primary
Antigen: IL17RA (interleukin 17 receptor A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR