Anti-ZKSCAN5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315268EA 552 EUR
ORIGTA315268
Anti-ZKSCAN5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: ZKSCAN5 (zinc finger with KRAB and SCAN domains 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR