Anti-ARHGEF12 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315241EA 552 EUR
ORIGTA315241
Anti-ARHGEF12 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: ARHGEF12 (Rho guanine nucleotide exchange factor (GEF) 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR