Anti-ZP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315202EA 552 EUR
ORIGTA315202
Anti-ZP1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ZP1 (zona pellucida glycoprotein 1 (sperm receptor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR