Anti-TAF6L Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315084EA 532 EUR
ORIGTA315084
Anti-TAF6L Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: TAF6L (TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR