Anti-TRIM16 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315083EA 532 EUR
ORIGTA315083
Anti-TRIM16 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: TRIM16 (tripartite motif containing 16)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR