Anti-KAT5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315069EA 552 EUR
ORIGTA315069
Anti-KAT5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: KAT5 (K(lysine) acetyltransferase 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR