Anti-SYNCRIP Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315062EA 564 EUR
ORIGTA315062
Anti-SYNCRIP Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: SYNCRIP (synaptotagmin binding, cytoplasmic RNA interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR