Anti-NMUR1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315043EA 552 EUR
ORIGTA315043
Anti-NMUR1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: NMUR1 (neuromedin U receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR