Anti-CST8 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315011EA 552 EUR
ORIGTA315011
Anti-CST8 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: CST8 (cystatin 8 (cystatin-related epididymal specific))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR