Anti-NCOR2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA314945EA 552 EUR
ORIGTA314945
Anti-NCOR2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:00-1:500; ELISA: 1:20000

Type: Primary
Antigen: NCOR2 (nuclear receptor corepressor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR