Anti-CDK5R2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA314841EA 552 EUR
ORIGTA314841
Anti-CDK5R2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: CDK5R2 (cyclin-dependent kinase 5, regulatory subunit 2 (p39))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR