Anti-SLC25A12 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA314778EA 552 EUR
ORIGTA314778
Anti-SLC25A12 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: SLC25A12 (solute carrier family 25 (aspartate/glutamate carrier), member 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR