Anti-PPP1R2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA314368EA 552 EUR
ORIGTA314368
Anti-PPP1R2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR