Anti-PDK1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303069EA 500 EUR
ORIGTA303069
Anti-PDK1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR