Anti-GYPA Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303062EA 547 EUR
ORIGTA303062
Anti-GYPA Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: GYPA (glycophorin A (MNS blood group))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR