Anti-CLU Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303058EA 500 EUR
ORIGTA303058
Anti-CLU Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: CLU (clusterin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR