Anti-STX1A / STX1B2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303051EA 500 EUR
ORIGTA303051
Anti-STX1A / STX1B2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: STX1A / STX1B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR