Anti-VDR Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303037EA 500 EUR
ORIGTA303037
Anti-VDR Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: VDR (vitamin D (1,25- dihydroxyvitamin D3) receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR