Anti-HTR2C Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303035EA 547 EUR
ORIGTA303035
Anti-HTR2C Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: HTR2C (5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR