Anti-NGFR Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA303032EA 500 EUR
ORIGTA303032
Anti-NGFR Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000 Western Blot: 03-1µg/ml

Type: Primary
Antigen: NGFR (nerve growth factor receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR