Anti-CHAT Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302990EA 547 EUR
ORIGTA302990
Anti-CHAT Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-03µg/ml

Type: Primary
Antigen: CHAT (choline O-acetyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR