Anti-MBL2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302979EA 451 EUR
ORIGTA302979
Anti-MBL2 Goat Polyclonal Antibody
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MBL2 (mannose-binding lectin (protein C) 2, soluble)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR