Anti-EFHD2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302973EA 502 EUR
ORIGTA302973
Anti-EFHD2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: EFHD2 (EF-hand domain family, member D2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR