Anti-BCL2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302955EA 500 EUR
ORIGTA302955
Anti-BCL2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR