Anti-MCL1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302945EA 500 EUR
ORIGTA302945
Anti-MCL1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MCL1 (myeloid cell leukemia 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR