Anti-PDLIM4 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302919EA 550 EUR
ORIGTA302919
Anti-PDLIM4 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000. Western Blot: 0.1-0.5µg/ml

Type: Primary
Antigen: PDLIM4 (PDZ and LIM domain 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR