Anti-NBEA Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302902EA 547 EUR
ORIGTA302902
Anti-NBEA Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: NBEA (neurobeachin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR