Anti-ARHGAP26 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302899EA 500 EUR
ORIGTA302899
Anti-ARHGAP26 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: ARHGAP26 (Rho GTPase activating protein 26)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR