Anti-PAFAH1B1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302796EA 500 EUR
ORIGTA302796
Anti-PAFAH1B1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: PAFAH1B1 (platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR