Anti-CREB3L4 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302777EA 547 EUR
ORIGTA302777
Anti-CREB3L4 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml; Immunochemistry: 2-4µg/ml

Type: Primary
Antigen: CREB3L4 (cAMP responsive element binding protein 3-like 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR