Anti-FRAT2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302774EA 550 EUR
ORIGTA302774
Anti-FRAT2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: FRAT2 (frequently rearranged in advanced T-cell lymphomas 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR