Anti-TMX1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302718EA 500 EUR
ORIGTA302718
Anti-TMX1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: TMX1 (thioredoxin-related transmembrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR