Anti-RAB2A Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302706EA 454 EUR
ORIGTA302706
Anti-RAB2A Goat Polyclonal Antibody
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: RAB2A (RAB2A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR