Anti-SOD1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302692EA 500 EUR
ORIGTA302692
Anti-SOD1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:8000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: SOD1 (superoxide dismutase 1, soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR