Anti-PLAU Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302671EA 451 EUR
ORIGTA302671
Anti-PLAU Goat Polyclonal Antibody
Recommended Dilutions: ELISA: 1:8000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: PLAU (plasminogen activator, urokinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR